Native Cmv antigen
Leverandør: ENZO LIFE SCIENCES
ALX6301250100
ENZOALX6301250100EA 7600 NOK
ENZOALX6301250100
Native Cmv antigen
Proteiner
From cytomegalovirus, strain AD169. Inactivated by 0.2% Triton X-100.