Anti-alpha Gal Epitope Mouse Monoclonal Antibody

Leverandør: ENZO LIFE SCIENCES
ALX8010901
ENZOALX8010901EA 7600 NOK
ENZOALX8010901
Anti-alpha Gal Epitope Mouse Monoclonal Antibody
Antistoffer
Host: Mouse, Isotype: IgM

  • For ELISA, Flow Cytometry, Immunohistochemistry, Western Blot

Recognizes synthetic and naturally produced mouse, rat and pig Galalpha1-3Gal epitopes on glycoproteins and glycolipids. Does not cross-react with beta-Gal glycoproteins or BSA.

Type: Primary
Antigen: alpha Gal Epitope
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR