Anti-CAMK2 Mouse Monoclonal Antibody

Leverandør: ENZO LIFE SCIENCES
ALX802009R100
ENZOALX802009R100EA 8025 NOK
ENZOALX802009R100
Anti-CAMK2 Mouse Monoclonal Antibody
Antistoffer
Host: Mouse, Isotype: IgG1

  • For ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot

Recognizes recombinant and native mouse and rat phosphorylated alpha-subunit of rat CaM kinase II.

Type: Primary
Antigen: CAMK2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR