Anti-NF-ATC1 Mouse Monoclonal Antibody

Leverandør: ENZO LIFE SCIENCES
ALX804022R100
ENZOALX804022R100EA 9875 NOK
ENZOALX804022R100
Anti-NF-ATC1 Mouse Monoclonal Antibody
Antistoffer
Host: Mouse, Isotype: IgG1

  • For Immunocytochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse, rat and monkey NF-ATc1. Detects a band of ~105kDa by Western blot. Does not cross-react with NF-ATc2.

Type: Primary
Antigen: NF-ATC1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR