Anti-EGFR Mouse Monoclonal Antibody

Leverandør: ENZO LIFE SCIENCES
ALX804064C100
ENZOALX804064C100EA 7400 NOK
ENZOALX804064C100
Anti-EGFR Mouse Monoclonal Antibody
Antistoffer
Host: Mouse, Isotype: IgG1

  • For ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot, Luminex

Recognizes the C-terminus of human and mouse EGF receptor (aa 1165-1186). Recognition is independent of phosphorylation of Tyr1173.

Reconstitute with 1ml water (15 minutes at room temperature).

Type: Primary
Antigen: EGFR
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
New Product
Bestill nå

SPESIFIKASJONER

Antigensymbol EGFR
Antigen-navn Epidermal growth factor receptor
Environmentally Preferable
Antigen synonymer PIG61|HER1|epidermal growth f|mENA|proto-oncogene c-ErbB-1|ERBB|avian erythroblastic leukemia viral (v-erb-b) oncogene homolog|cell proliferation-inducing protein 61|cell growth inhibiting protein 40|receptor tyrosine-protein kinase erbB-1|ERBB1
Antistofftype Primary
Klonalitet Monoclonal
Reaktivitet Human
Vert Mouse
Isotype IgG1
Western blot Yes
ELISA Yes
ImmunoKjemi Yes
Immunutfelling Yes
Skjema Lyophilised from 1 ml of 2X PBS containing 0,09% sodium azide, PEG, and sucrose.
Rensing Thiophilic adsorption and size exclusion chromatography purified

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR