Anti-GRP75 Mouse Monoclonal Antibody

Leverandør: ENZO LIFE SCIENCES
ALX804077R100
ENZOALX804077R100EA 9875 NOK
ENZOALX804077R100
Anti-GRP75 Mouse Monoclonal Antibody
Antistoffer
Host: Mouse, Isotype: IgG3

  • For Immunocytochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse, dog and monkey mtHSP70. Detects a band of ~75kDa by Western blot.

Type: Primary
Antigen: GRP75
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
New Product
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR