Anti-APMAP Mouse Monoclonal Antibody

Leverandør: ENZO LIFE SCIENCES
ALX804144C100
ENZOALX804144C100EA 5850 NOK
ENZOALX804144C100
Anti-APMAP Mouse Monoclonal Antibody
Antistoffer
Host: Mouse, Isotype: IgG1

  • For ELISA, Flow Cytometry, Immunoprecipitation, Western Blot

Recognizes human ACRP30headless. Does not cross-react with mouse ACRP30headless. Does not cross-react with full length adiponectin.

Type: Primary
Antigen: APMAP
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR