Anti-BCMA Rat Monoclonal Antibody [clone: Vicky-1]

Leverandør: ENZO LIFE SCIENCES
ALX804151C100
ENZOALX804151C100EA 6650 NOK
ENZOALX804151C100
Anti-BCMA Rat Monoclonal Antibody [clone: Vicky-1]
Antistoffer
BCMA (B cell maturation) is a nonglycosylated integral membrane type I protein that is preferentially expressed in mature B lymphocytes and belongs structurally to the TNF receptor family. BCMA associates with BAFF, APRIL, TNF receptor-associated factor (TRAF) 1, TRAF2, and TRAF3 and activates NF-κB, Elk-1, c-Jun N-terminal kinase (JNK), and p38 mitogen-activated protein kinase. BCMA plays an important role in B cell development, function and regulation.

Type: Primary
Antigen: BCMA
Clonality: Monoclonal
Clone: Vicky-1
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR