Anti-HBVX Mouse Monoclonal Antibody

Leverandør: ENZO LIFE SCIENCES
ALX804278C100
ENZOALX804278C100EA 10400 NOK
ENZOALX804278C100
Anti-HBVX Mouse Monoclonal Antibody
Antistoffer
Host: Mouse, Isotype: IgG1

  • For ELISA, Flow Cytometry, Immunoprecipitation, Western Blot

Recognizes HBx. Detects a band of ~17kDa by Western blot.

Type: Primary
Antigen: HEPATITIS BX PROTEIN
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Virus
New Product
Bestill nå

SPESIFIKASJONER

Antigensymbol HBVX
Environmentally Preferable
Antigen-navn Hepatitis B Virus X Protein
Antistofftype Primary
Klonalitet Monoclonal
Reaktivitet Virus
Vert Mouse
Isotype IgG1
Western blot Yes
ELISA Yes
Immunutfelling Yes
Skjema Liquid. In PBS containing 0,05% sodium azide
Konsentrasjon 1 mg/ml

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR