Anti-BMF Rat Monoclonal Antibody

Leverandør: ENZO LIFE SCIENCES
ALX804343C100
ENZOALX804343C100EA 6425 NOK
ENZOALX804343C100
Anti-BMF Rat Monoclonal Antibody
Antistoffer
Host: Rat, Isotype: IgG2a

  • For Immunoprecipitation, Western Blot

Recognizes human and mouse Bmf.

Type: Primary
Antigen: BMF
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR