Anti-AKT Mouse Monoclonal Antibody

Leverandør: ENZO LIFE SCIENCES
ALX804401C100
ENZOALX804401C100EA 8475 NOK
ENZOALX804401C100
Anti-AKT Mouse Monoclonal Antibody
Antistoffer
Host: Mouse, Isotype: IgG1

  • For ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse, rat and dog Ser472/473/474-phosphorylated Akt (see technical note). Does not cross-react with dephosphorylated Akt or other unrelated serine-phosphorylated proteins.

Reconstitute with 1ml water (15 minutes at room temperature).

Type: Primary
Antigen: AKT
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
New Product
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR