Anti-Tubulin Beta-3 Mouse Monoclonal Antibody

Leverandør: ENZO LIFE SCIENCES
ALX804405C100
ENZOALX804405C100EA 3775 NOK
ENZOALX804405C100
Anti-Tubulin Beta-3 Mouse Monoclonal Antibody
Antistoffer
Host: Mouse, Isotype: IgG1

  • For ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot

Recognizes human betaIII-tubulin located in neurons.

Type: Primary
Antigen: TUBULIN BETA-3
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR