Anti-SYK Mouse Monoclonal Antibody

Leverandør: ENZO LIFE SCIENCES
ALX804480C100
ENZOALX804480C100EA 4225 NOK
ENZOALX804480C100
Anti-SYK Mouse Monoclonal Antibody
Antistoffer
Host: Mouse, Isotype: IgG1

  • For Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse and rat Syk.

Type: Primary
Antigen: SYK
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR