Anti-BIMS/EL/L Rat Monoclonal Antibody

Leverandør: ENZO LIFE SCIENCES
ALX804527C100
ENZOALX804527C100EA 6250 NOK
ENZOALX804527C100
Anti-BIMS/EL/L Rat Monoclonal Antibody
Antistoffer
Host: Rat, Isotype: IgG2a

  • For ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot

Recognizes an epitope within aa 20-40 of human, mouse, rat, monkey and dog BimL (~19kDa), BimEL (~23kDa), and BimS (~16kDa). Also recognizes the six novel isoforms of human Bim, generated by alternative splicing and lacking the C-terminal hydrophobic region recently identified.

Type: Primary
Antigen: BIMS/EL/L
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR