Anti-PIDD Mouse Monoclonal Antibody

Leverandør: ENZO LIFE SCIENCES
ALX804837C100
ENZOALX804837C100EA 5900 NOK
ENZOALX804837C100
Anti-PIDD Mouse Monoclonal Antibody
Antistoffer
Host: Mouse, Isotype: IgG1

  • For Immunohistochemistry, Western Blot

Recognizes human and mouse PIDD. Detects bands of ~90kDa (full length protein) and ~50kDa (cleaved protein) by Western blot.

Type: Primary
Antigen: PIDD
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR