Anti-MHC1 Mouse Monoclonal Antibody

Leverandør: ENZO LIFE SCIENCES
ALX805711C100
ENZOALX805711C100EA 5275 NOK
ENZOALX805711C100
Anti-MHC1 Mouse Monoclonal Antibody
Antistoffer
HLA-class I major histocompatibility (MHC) antigens are intrinsic membrane glycoproteins expressed on nucleated cells and noncovalently associated with an invariant beta2 microglobulin. They carry foreign determinants important for immune recognition by cytotoxic T cells, thus important for anti-viral and anti-tumour defence. Human HLA-class I antigens are represented by HLA-A, HLA-B and HLA-C molecules.

  • For ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human, monkey, bovine and cat MHCMore Product Information
New Product
Bestill nå

SPESIFIKASJONER

Antigensymbol MHC1
Antigen-navn MHC Class I
Environmentally Preferable
Antistofftype Primary
Klonalitet Monoclonal
Reaktivitet
Vert Mouse
Isotype IgG2a
Western blot Yes
ELISA Yes
ImmunoKjemi Yes
Immunutfelling Yes
Flowcytometri Yes
Skjema Liquid. In PBS, pH 7,4. Sterile filtered. Contains no preservatives
Rensing ≥95% (SDS-PAGE)
Konsentrasjon 1 mg/ml

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR