DNA (Deoxyribonucleic acid) (from calf thymus), purified

Leverandør: ENZO LIFE SCIENCES

ALX840040C010
ENZOALX840040C010EA 1300 NOK
ENZOALX840040C010
DNA (Deoxyribonucleic acid) (from calf thymus), purified
DNA (Deoxyribonucleic acid)

Purified from calf thymus and activated by digestion with DNAse I.


For use with PARP-1 or PARP-2 in vitro activity tests


Concentration: 0,2 mg/ml

Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00130922
CAS-nummer: 9007-49-2

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR