Human recombinant TRAIL, soluble
Leverandør: ENZO LIFE SCIENCES
ALX850018KI01
ENZOALX850018KI01EA 9825 NOK
ENZOALX850018KI01
Human recombinant TRAIL, soluble
Proteiner
Induces apoptosis in a concentration range of 1 to100 ng/ml if applied with the cross-linking enhancer.
Bestillingsinformasjon: Contents: 10 µg of TRAIL, Soluble (human) (recombinant) (Prod. No. ALX-522-003) and 2×50 µg of Enhancer for Ligands (Prod. No. ALX-804-034).