RANKL (total), soluble (human) ELISA kit
Leverandør: ENZO LIFE SCIENCES
ALX850019KI01
ENZOALX850019KI01EA 15575 NOK
ENZOALX850019KI01
RANKL (total), soluble (human) ELISA kit
Assay ELISAs
For the quantitative determination of free and OPG-bound human soluble RANKL from serum, plasma and cell culture supernatant.
New Product
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Primary antibody reactivity Human
Vert
Sensitivitet ~1,56 pg/ml (range 2,2 - 60 pmol/L)