ApoSENSOR™ Cell viability assay kit
Leverandør: ENZO LIFE SCIENCES
ALX850247KI01
ENZOALX850247KI01EA 4250 NOK
ENZOALX850247KI01
ApoSENSOR™ Cell viability assay kit
Assay Cellulær analyse
The assay utilizes bioluminescent detection of the ATP level for a rapid screening of apoptosis and cell proliferation simultaneously in mammalian cells. The assay utilizes the enzyme luciferase to catalyze the formation of light from ATP and luciferin. The light can be measured using a luminometer or Beta Counter. The assay can be fully automatic for high throughput (10 seconds/sample) and is highly sensitive (detects 10-100 mammalian cells/well).
Leveringsinformasjon: Kit includes Nucleotide Releasing Buffer, ATP Monitoring Enzyme, Enzyme Reconstitution Buffer and ATP.