PTX3 (human) detection set [for ELISA application]
Leverandør: ENZO LIFE SCIENCES
ALX850299KI01
ENZOALX850299KI01EA 15700 NOK
ENZOALX850299KI01
PTX3 (human) detection set [for ELISA application]
Assay ELISAs
For the quantitative determination of PTX3 in human and bovine (weak) cell culture media, EDTA-plasma or serum (weak) samples.
Leveringsinformasjon: Kit includes 1 vial (10 µg) PTX3 (human), 1 vial (50 µg) PTX3, pAb (Biotin), 1 vial (100 µg) PTX3, mAb (MNB4) for preparing 10 plates.
New Product
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Mål PTX3
Primary antibody reactivity Human
Vert
Lagringsforhold −20 °C (long term)