LBP, soluble (mouse) ELISA kit
Leverandør: ENZO LIFE SCIENCES
ALX850305KI01
ENZOALX850305KI01EA 13925 NOK
ENZOALX850305KI01
LBP, soluble (mouse) ELISA kit
Assay ELISAs
For the quantitative determination of mouse and rat soluble as well as bound LBP levels in serum, plasma and cell culture medium.
Bestillingsinformasjon: Contains 1 precoated ELISA module1 vial anti-LBP (mouse) detecting MAb1 vial LBP standard (mouse)1 vial reference serum (mouse)2 tablets PBS1 vial dilution buffer1 vial Tween 201 vial stopping solution1 vial substrate solution.