Microcystins (adda specific) ELISA kit
Leverandør: ENZO LIFE SCIENCES
ALX850319KI01
ENZOALX850319KI01EA 12300 NOK
ENZOALX850319KI01
Microcystins (adda specific) ELISA kit
Assay ELISAs
Polyclonal antibody prepared against the Adda moiety binds to microcystins and nodularins, allowing the congener-independent determination of these toxins and many of its congeners, and does not cross-react with other non-related toxins or compounds. No pre-sample preparation required. Enables simultaneous measurement of multiple samples at reasonable costs.
  • Convenient – no pre-sample preparation required
  • Rapid – analyze 41 duplicate samples in <2,5 hours
  • Assay Time: 1 hour 90 minutes
New Product
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Primary antibody reactivity
Vert
Sensitivitet 0,1 ng/ml (Range 0,15 - 5 ng/ml)
Lagringsforhold 4 °C (long term)