Orphan ligand library plates, SCREEN-WELL®
Leverandør: ENZO LIFE SCIENCES
BML28250500
ENZOBML28250500EA 34525 NOK
ENZOBML28250500
Orphan ligand library plates, SCREEN-WELL®
Mikroplater HTS-mikroplater
SCREEN-WELL® orphan ligand library contains 84 compounds with defined, putative, potential or speculative biological activity but whose protein binding partners have not been identified.
The library is a rich source of ligands for receptor de-orphaning. Includes the following classes of ligands: Trace Amines, Neurotransmitter Metabolites, Endogenous β-carbolines, Urinary Metabolites, Nicotine Congeners, D-Amino Acids. Receptors for endogenous trace amines have been deorphanized. Endogenous β-carbolines have been shown to display varied biological effects such as hallucinations,More Product Information