ICCB known bioactives library plates, SCREEN-WELL®
Leverandør: ENZO LIFE SCIENCES
BML28400100
ENZOBML28400100EA 216700 NOK
ENZOBML28400100
ICCB known bioactives library plates, SCREEN-WELL®
Mikroplater HTS-mikroplater
SCREEN-WELL® ICCB Known Bioactives Library is a collection of 472 diverse biologically active compounds with defined biological activity and was developed in collaboration with the Harvard Institute of Chemistry and Cell Biology.
SCREEN-WELL® ICCB Known Bioactives Library includes the following classes of compounds: GPCR ligands, Second messenger modulators, Nuclear receptor ligands, Actin and tubulin modulators, Kinase inhibitors, Protease inhibitors, Ion channel blockers, Gene regulation agents, Lipid biosynthesis inhibitors, Phosphodiesterase inhibitors, and many other classes. EachMore Product Information