Puromycin aminonucleoside ≥98% (by TLC)

Leverandør: ENZO LIFE SCIENCES

BMLA2600050
ENZOBMLA2600050EA 3275 NOK
ENZOBMLA2600050
Puromycin aminonucleoside ≥98% (by TLC)
Puromycin aminonucleoside

A puromycin analog which does not inhibit protein synthesis or induce apoptosis. It is a glomerular epithelial cell toxin which may be used to induce nephropathy in laboratory animals.


  • White solid


Soluble in water (10 mg/ml)


Forpakning: Shipping temperature: Ambient

Formel: C₁₂H₁₈N₆O₃
Smeltepunkt: 222…223 °C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00063462
CAS-nummer: 58-60-6

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR