CV-Cathepsin B detection kit
Leverandør: ENZO LIFE SCIENCES
BMLAK1250001
ENZOBMLAK1250001EA 4450 NOK
ENZOBMLAK1250001
CV-Cathepsin B detection kit
Assay Cellulær analyse
Based on the dye cresyl violet and the cathepsin B ArgArg peptide recognition sequence, this kit allows the detection of cathepsin B activity in living cells by fluorescence microscopy.
Once cells are grown, staining is completed in just a few minutes. Hoechst 33342 stain and Acridine Orange are also included to allow visualization of nuclei and lysosomes.
Elevated cathepsin enzyme activity often indicates the presence of a pathological condition including cancer, Alzheimer's disease, autoimmune disease, and osteoporosis.
Leveringsinformasjon: Kit contains CV-(RR)2 reagent, Acridine Orange, Hoechst 33342 stain
New Product
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Applikasjon Fluorescence microscopy, fluorescent detection
Størrelse 1 kit
Lagringsforhold 4 °C