ALDetect™ (MDA-Specific) Lipid Peroxidation Assay kit
Leverandør: ENZO LIFE SCIENCES
BMLAK1710001
ENZOBMLAK1710001EA 9900 NOK
ENZOBMLAK1710001
ALDetect™ (MDA-Specific) Lipid Peroxidation Assay kit
Assay Cellulær analyse
Lipid peroxidation is a well-established mechanism of cellular injury in both plants and animals, and is used as an indicator of oxidative stress in cells and tissues. Lipid peroxides, derived from polyunsaturated fatty acids, are unstable and decompose to form a complex series of compounds. These include reactive aldehydes, of which the most abundant is malondialdehyde (MDA). Therefore, measurement of malondialdehyde is widely used as an indicator of lipid peroxidation. Increased levels of lipid peroxidation products have been associated with a variety of chronic diseases in both humans and model systems.
MDA reacts readily with amino groups on proteins and other biomolecules to form a variety of adducts, including cross-linked products. MDA also forms adducts with DNA bases that are mutagenic and possibly carcinogenic. DNA-protein cross-links are another result of the reaction between DNA and MDA.
The TBARS method is commonly used to measure MDA in biological samples. However, this reaction is relatively nonspecific; both free and protein-bound MDA can react. The method is designed to assay free MDA or, after a hydrolysis step, total MDA (i.e., free and protein-bound Schiff base conjugates). The assay conditions serve to minimize interference from other lipid peroxidation products, such as 4-hydroxyalkenals
Leveringsinformasjon: Kit includes Reagent R1: N-methyl-2-phenylindole, in acetonitrile (3×18 ml), Reagent R2: Conc. Hydrochloric acid (1×16,5 ml), MDA Standard: 1,1,3,3-tetramethoxypropane (TMOP) in Tris-hydrochloric acid (1 ml), BHT Butylated hydroxytoluene in acetonitrile (2 ml), Probucol in methanol (1,1 ml), Methanol (30 ml).
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Applikasjon Colorimetric detection
Størrelse 25 tests
Lagringsforhold 4 °C (long term)