Matrix Metalloproteinase-3 (MMP-3) colorimetric drug discovery kit

Leverandør: ENZO LIFE SCIENCES

BMLAK4000001
ENZOBMLAK4000001EA 7800 NOK
ENZOBMLAK4000001
Matrix Metalloproteinase-3 (MMP-3) colorimetric drug discovery kit
Assay Enzymanalyser

A QuantiZyme™ Assay System. The MMP-3 Colorimetric Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-3 (MMP-3, stromelysin-1) using a thiopeptide as a chromogenic substrate. The assays are performed in a convenient 96 well microplate format.


Leveringsinformasjon: Kit contains MMP-3 Enzyme (human) (recombinant), Substrate, Inhibitor, Assay Buffer and 96-well Microplate.

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR