Matrix Metalloproteinase-3 (MMP-3) fluorometric drug discovery kit

Leverandør: ENZO LIFE SCIENCES

BMLAK4010001
ENZOBMLAK4010001EA 7400 NOK
ENZOBMLAK4010001
Matrix Metalloproteinase-3 (MMP-3) fluorometric drug discovery kit
Assay Enzymanalyser

A QuantiZyme™ Assay System. The MMP-3 Fluorometric (also known as fluorimetric) Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-3 (MMP-3, stromelysin-1) using a quenched fluorogenic peptide: OmniMMP™ fluorogenic substrate Mca-Pro-Leu-Gly-Leu-Dpa-Ala-Arg-NH. Mca fluorescence is quenched by the Dpa group until cleavage by MMPs at the Gly-Leu bond separates the two moieties.


Leveringsinformasjon: Kit contains MMP-3 Enzyme (human) (recombinant), Substrate, Calibration Standard, Prototypic Control Inhibitor (NNGH3), Assay Buffer, 96-well White Microplate and a detailed instruction booklet.

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR