Matrix Metalloproteinase-1 (MMP-1) colorimetric drug discovery kit

Leverandør: ENZO LIFE SCIENCES

BMLAK4040001
ENZOBMLAK4040001EA 7275 NOK
ENZOBMLAK4040001
Matrix Metalloproteinase-1 (MMP-1) colorimetric drug discovery kit
Assay Enzymanalyser

A QuantiZyme™ Assay System. The MMP-1 Colorimetric Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-1 (MMP-1, interstitial collagenase, fibroblast collagenase) using a thiopeptide as a chromogenic substrate.


Leveringsinformasjon: Kit includes MMP-1 Enzyme (human) (recombinant), Substrate, inhibitor, Assay Buffer and 96-well Microplate.

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR