Matrix Metalloproteinase-9 (MMP-9) colorimetric drug discovery kit

Leverandør: ENZO LIFE SCIENCES

BMLAK4100001
ENZOBMLAK4100001EA 7800 NOK
ENZOBMLAK4100001
Matrix Metalloproteinase-9 (MMP-9) colorimetric drug discovery kit
Assay Enzymanalyser

A QuantiZyme™ Assay System. The MMP-9 Colorimetric Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-9 (MMP-9, gelatinase B) using a thiopeptide as a chromogenic substrate.


Leveringsinformasjon: Kit contains MMP-9 Enzyme (human) (recombinant), Substrate, Calibration Standard, Prototypic Control Inhibitor (NNGH), Assay Buffer, 96-well White Microplate and a detailed instruction booklet.

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR