Matrix Metalloproteinase-9 (MMP-9) fluorometric drug discovery kit

Leverandør: ENZO LIFE SCIENCES

BMLAK4110001
ENZOBMLAK4110001EA 7400 NOK
ENZOBMLAK4110001
Matrix Metalloproteinase-9 (MMP-9) fluorometric drug discovery kit
Assay Enzymanalyser

A QuantiZyme™ Assay SystemThe MMP-9 Fluorometric (also known as fluorimetric) Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-9 (MMP-9, gelatinase B) using a quenched fluorogenic peptide: OmniMMP™ fluorogenic substrate Mca-Pro-Leu-Gly-Leu-Dpa-Ala-Arg-NH. Mca fluorescence is quenched by the Dpa group until cleavage by MMPs at the Gly-Leu bond separates the two moieties.


Leveringsinformasjon: Kit contains MMP-9 Enzyme (human) (recombinant), Substrate, Calibration Standard, Prototypic Control Inhibitor (NNGH3), Assay Buffer, 96-well White Microplate and a detailed instruction booklet.

New Product

Bestill nå

SPESIFIKASJONER

Applikasjon Fluorescent detection, HTS activity assay
Environmentally Preferable
Lagringsforhold −80 °C
Størrelse 1 kit

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR