Matrix Metalloproteinase-13 (MMP-13) fluorometric drug discovery kit
Leverandør: ENZO LIFE SCIENCES
BMLAK4130001
ENZOBMLAK4130001EA 7875 NOK
ENZOBMLAK4130001
Matrix Metalloproteinase-13 (MMP-13) fluorometric drug discovery kit
Assay Enzymanalyser
A QuantiZyme™ Assay System. The MMP-13 Fluorometric (also known as fluorimetric) Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-13 (MMP-13, collagenase-3) using a quenched fluorogenic peptide: OmniMMP™ fluorogenic substrate Mca-Pro-Leu-Gly-Leu-Dpa-Ala-Arg-NH. Mca fluorescence is quenched by the Dpa group until cleavage by MMPs at the Gly-Leu bond separates the two moieties.
Leveringsinformasjon: Kit contains MMP-13 Enzyme (human) (recombinant), Substrate, Calibration Standard, Prototypic Control Inhibitor (NNGH3), Assay Buffer, 96-well White Microplate and a detailed instruction booklet.
New Product