Matrix Metalloproteinase-8 (MMP-8) colorimetric drug discovery kit

Leverandør: ENZO LIFE SCIENCES

BMLAK4140001
ENZOBMLAK4140001EA 7425 NOK
ENZOBMLAK4140001
Matrix Metalloproteinase-8 (MMP-8) colorimetric drug discovery kit
Assay Enzymanalyser

A QuantiZyme™ Assay System. The MMP-8 Colorimetric Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-8 (MMP-8, neutrophil collagenase, collagenase-2) using a thiopeptide as a chromogenic substrate.


Leveringsinformasjon: Kit contains MMP-8 Enzyme (human) (recombinant), Substrate,, Prototypic Control Inhibitor (NNGH3), Assay Buffer, 96-well White Microplate and a detailed instruction booklet.

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR