Matrix Metalloproteinase-14 (MMP-14) colorimetric drug discovery kit

Leverandør: ENZO LIFE SCIENCES

BMLAK4160001
ENZOBMLAK4160001EA 7400 NOK
ENZOBMLAK4160001
Matrix Metalloproteinase-14 (MMP-14) colorimetric drug discovery kit
Assay Enzymanalyser

A QuantiZyme™ Assay System. The MMP-14 Colorimetric Drug Discovery Kit is a complete assay sytem designed to screen for inhibitors of matrix metalloproteinase-14 (MMP-14, membrane-type matrix metalloproteinase 1, MT1-MMP) using a thiopeptide as a chromogenic substrate.


Leveringsinformasjon: Kit contains MMP-14 Enzyme (human) (recombinant), Substrate, Calibration Standard, Prototypic Control Inhibitor (NNGH), Assay Buffer, 96-well White Microplate and a detailed instruction booklet.

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR