Cathepsin K drug discovery kit
Leverandør: ENZO LIFE SCIENCES
BMLAK4300001
ENZOBMLAK4300001EA 7150 NOK
ENZOBMLAK4300001
Cathepsin K drug discovery kit
Assay Enzymanalyser
The Cathepsin K Drug Discovery Kit is a complete assay system designed to screen inhibitors of cathepsin K. Using a convenient microplate format, cathepsin K activity is monitored fluorimetrically using the substrate provided in the kit.
Bestillingsinformasjon: Quantity: 96 assays Contents: Cathepsin K (human) (recombinant), AMC substrate, AMC calibration standard, inhibitor, assay buffer, ½-volume white microplate.