Calcineurin phosphatase assay kit
Leverandør: ENZO LIFE SCIENCES
BMLAK8040001
ENZOBMLAK8040001EA 5900 NOK
ENZOBMLAK8040001
Calcineurin phosphatase assay kit
Assay Enzymanalyser
This colorimetric kit is designed for inhibitor screening or kinetic assay of recombinant Calcineurin. Activity is determined using the RII phosphopeptide substrate, the most efficient and selective peptide known for calcineurin, and detection of free phosphate released is based on the classic malachite green assay.
  • Safe and non-radioactive assay
  • Convenient 1-step detection, no mixing needed
Leveringsinformasjon: Kit includes Recombinant calcineurin, RII phosphopeptide substrate, calmodulin, assay buffer, BIOMOL® GREEN detection reagent,
phosphate standard, microplate and detailed instructions.