Carvedilol ≥98% (by HPLC)

Leverandør: ENZO LIFE SCIENCES
Warning

Synonymer: 1-(9H-Carbazol-4-yloxy)-3-[[2-(2-methoxyphenoxy)ethyl]amino]-2-propanol

BMLAR1120100
ENZOBMLAR1120100EA 995 NOK
ENZOBMLAR1120100
Carvedilol ≥98% (by HPLC)
Carvedilol

Adrenergic receptor ligand.


An antagonist at α1- and β-adrenergic receptors. Displays antioxidant and antiapoptotic effects in vivo.


Soluble in DMSO (>25 mg/ml) or 100% ethanol (10 mg/ml).

Formel: C₂₄H₂₆N₂O₄
MW: 406,48 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00864692
CAS-nummer: 72956-09-3
FN: 3077
ADR: 9,III

Bestill nå

Spesifikasjon Testresultater

Purity ≥98% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR