BAPTA-AM ≥90% (by TLC)

Leverandør: ENZO LIFE SCIENCES

Synonymer: Tetrakis(acetoxymethyl)-1,2-bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetate, Tetrakis(acetoxymethyl) 1,2-Bis(2-aminophenoxy)ethane-N,N,N,N-tetraacetate, 1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic Acid Tetrakis(acetoxymethyl) Ester, BAPTA-AM ester

BMLCA4110025
ENZOBMLCA4110025EA 2750 NOK
ENZOBMLCA4110025
BAPTA-AM ≥90% (by TLC)
BAPTA-AM

PKC activator


BAPTA is a Ca²⁺ chelator with 105-fold greater affinity for Ca²⁺ than for Mg2+. BAPTA-AM is a cell-permeable derivative which is widely used as an intracellular calcium sponge. Induces inactivation of protein kinase C. Also inhibits thapsigargin-induced apoptosis in rat thymocytes.

Formel: C₃₄H₄₀N₂O₁₈
MW: 764.69 g/mol
MDL nummer: MFCD00036696
CAS-nummer: 126150-97-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR