8-(4-Chlorophenylthio)guanosine 3',5'-cyclic Monophosphothioate, Rp-Isomer triethylammonium salt ≥98% (by HPLC)

Leverandør: ENZO LIFE SCIENCES

BMLCN2060001
ENZOBMLCN2060001EA 4100 NOK
ENZOBMLCN2060001
8-(4-Chlorophenylthio)guanosine 3',5'-cyclic Monophosphothioate, Rp-Isomer triethylammonium salt ≥98% (by HPLC)
8-(4-Chlorophenylthio)guanosine 3',5'-cyclic Monophosphothioate, Rp-Isomer triethylammonium salt

A cell-permeable cGMP antagonist which is resistant to hydrolysis by cyclic nucleotide phosphodiesterases (Ki=0.5 µM). No metabolic side effects. Significantly more lipophilic and membrane permeant as compared to Rp-cGMPS or Rp-8-Br-cGMPS


  • PKG inhibitor


White solid.


Soluble in water (25 mg/ml).

Formel: C₁₆H₁₄ClN₅O₆PS₂·C₆H₁₆N
Lagringstemperatur: Fryseskap
CAS-nummer: 153660-04-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f8c64d23384b3e6b33342015b53a13e7022bbbd&vid=89c2ebae85c1593023f2cde1d9ee0a3390595273&prid=web&clid=SAPDAM
New Product

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR