AM-404 98 TLC_ASSAY_METHOD

Leverandør: ENZO LIFE SCIENCES

BMLCR1060050
ENZOBMLCR1060050EA 3800 NOK
ENZOBMLCR1060050
AM-404 98 TLC_ASSAY_METHOD
AM-404

Anandamide transport inhibitor


Analog of anandamide (Prod. No. BML-FA017). Potentiates the activity of endogenous anandamide by blocking its re-uptake into presynaptic membranes. Activates TRPV1 (EC50=0.04µM) at concentrations lower than those required to inhibit anandamide transport into the cell (neuronal (C6 glioma cells): IC50=10µM; non-neuronal (rat RBL-2H3 cells): IC50=11µM). Low affinity to FAAH (IC50=5.9µM), to CB1 receptor (IC50=1.76µM) and to CB2 receptor (IC50>1µM).

Formel: C₂₆H₃₇NO₂
MW: 395.587 g/mol
MDL nummer: MFCD03095721
CAS-nummer: 198022-70-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7793c6931c987aa133e1e25e402fe41560f9dc8c&vid=0e70b2ed496d3ce26460bfdd8e6d9b107bde38f4&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR