AM-251 ≥98% (by HPLC)

Leverandør: ENZO LIFE SCIENCES

Synonymer: 1-(2,4-Dichlorophenyl)-5-(4-iodophenyl)-4-methyl-N-1-piperidinyl-1H-pyrazole-3-carboxamide, 1-(2,4-Dichlorophenyl)-5-(4-iodophenyl)-4-methyl-N-(piperidin-1-yl)-1H-pyrazole-3-carboxamide, AM 251, AM251, AM-251

BMLCR1080010
ENZOBMLCR1080010EA 3600 NOK
ENZOBMLCR1080010
AM-251 ≥98% (by HPLC)
AM-251

Cannabinoid receptor ligand.


Cannabinoid CB1 receptor antagonist (Ki = 7.5 nM). Displays a high level of selectivity (306-fold) for CB1 over CB2 receptors. Structurally related to the cannabinoid receptor (CB) antagonist SR 141716A.


Soluble in DMSO, 100% ethanol or dimethyl formamide.

Formel: C₂₂H₂₁Cl₂IN₄O
MW: 555.2 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD01861181
CAS-nummer: 183232-66-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42a1a4548942669181ccd2432e454d6d1a130062&vid=ff397d38d11f971ff345f539e4a48c0c8c52e6d5&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Purity ≥98% (HPLC)
Identity Determined by ¹H-NMR and MS


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR