Clozapine ≥99% (by HPLC)

Leverandør: ENZO LIFE SCIENCES
Danger

Synonymer: 8-Chloro-11-(4-methyl-1-piperazinyl)-5H-dibenzo[b,e][1,4]diazepine

BMLD1070050 BMLD1070500
ENZOBMLD1070050EA 995 NOK
ENZOBMLD1070050 ENZOBMLD1070500
Clozapine ≥99% (by HPLC)
Clozapine

Dopamine receptor and serotonin receptor antagonist.


Dopamine D₄ and D₂ receptor antagonist. High affinity for the cloned rat dopamine D4 receptor (Ki<20 nM). Atypical neuroleptic agent. Antagonist at 5HT2A, 5HT2C, 5HT3, 5HT6 and 5HT7 receptors.


Soluble in DMSO (30 mg/ml) or dilute aqueous acid.

Formel: C₁₈H₁₉ClN₄
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00153785
CAS-nummer: 5786-21-0
EINECS: 227-313-7
FN: 2811
ADR: 6.1,III
Merck Indeks: 14,02423
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b6ce4b4b8154685056959613f75f535b469be6fb&vid=d958514cdd1f8ce5e7ea8598c9cb0d0a9f428755&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Purity ≥99% (HPLC)
Identity Determined by IR


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR