Oxatomide ≥99% (by HPLC)

Leverandør: ENZO LIFE SCIENCES

Synonymer: 1-(3-(4-Benzhydrylpiperazin-1-yl)propyl)-1,3-dihydro-2H-benzo[d]imidazol-2-one

BMLEI1140010
ENZOBMLEI1140010EA 1950 NOK
ENZOBMLEI1140010
Oxatomide ≥99% (by HPLC)
Oxatomide

Suppresses PAF-induced bronchoconstriction in the guinea pig (ED50=0.9 mg/kg orally).


  • Inhibits the release and actions of leukotrienes


White solid.


Soluble in ethanol or DMSO.

Formel: C₂₇H₃₀N₄O
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00211147
CAS-nummer: 60607-34-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5d5169ceb4e0f5c91497909ce6a76b223d2a9992&vid=394a6684e442727c5c46ae9ac64fc90b0918354e&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR