Tyrphostin A23 ≥99% (by TLC)

Leverandør: ENZO LIFE SCIENCES

BMLEI1910010
ENZOBMLEI1910010EA 1025 NOK
ENZOBMLEI1910010
Tyrphostin A23 ≥99% (by TLC)
Tyrphostin A23

A potent inhibitor of EGF receptor kinase activity (IC50=35 µM).


  • EGFR inhibitor


Yellow-tan solid.


Soluble in DMSO (25mg/ml) or 100% ethanol (25mg/ml).

Formel: C₁₀H₆N₂O₂
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00133899
CAS-nummer: 118409-57-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=10ff874f175636f144a66dbe59a73a1902dd9104&vid=00718d3fb1c3c059f9e97d1dc35d0dea2d0a69a2&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR