Tyrphostin A9 ≥99% (by HPLC)

Leverandør: ENZO LIFE SCIENCES

Synonymer: [4-Hydroxy-3,5-bis(2-methyl-2-propanyl)benzylidene]malononitrile, Malonoben

BMLEI2150050 BMLEI2150250
ENZOBMLEI2150050EA 1050 NOK
ENZOBMLEI2150050 ENZOBMLEI2150250
Tyrphostin A9 ≥99% (by HPLC)
Tyrphostin A9

Tyrphostin 9 is a selective inhibitor of the PDGF receptor tyrosine kinase (IC50=1.2 µM). It is also a potent (10 nM) uncoupler of oxidative phosphorylation.


  • PDGF receptor inhibitor


Yellow solid.


Soluble in DMSO (>25mg/ml) or ethanol (20mg/ml).

Formel: C₁₈H₂₂N₂O
MW: 282.387 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00209853
CAS-nummer: 10537-47-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b650322c94c0ed0e241e857bb9af473ad1740fe&vid=d5153d7da389d78435ad30c86565eb697f4d917f&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR