Benzylphosphonic acid-(AM)2 98 TLC_ASSAY_METHOD

Leverandør: ENZO LIFE SCIENCES

BMLEI2450010
ENZOBMLEI2450010EA 995 NOK
ENZOBMLEI2450010
Benzylphosphonic acid-(AM)2 98 TLC_ASSAY_METHOD
Benzylphosphonic acid-(AM)2

Benzylphosphonic acid-(AM)2 is a bis-acetoxymethyl ester (AM) derivative of benzylphosphonic acid which is cell permeable and is converted to the active phosphonic acid via intracellular esterase-catalyzed cleavage of the AM esters.


  • PTP inhibitor


White solid.


Soluble in ethanol (20 mg/ml) or DMSO (20 mg/ml).

Formel: C₁₃H₁₇O₇P
MW: 316.3 g/mol
Smeltepunkt: 38…39 °C
Lagringstemperatur: Fryseskap
CAS-nummer: 200716-68-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a73094c972fb6e20fb9acab07fea5ac35766ab7&vid=12fae46b41a291e814870d51c668267d67b78f0e&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR