Bisindolylmaleimide IX methanesulphonate ≥98%

Leverandør: ENZO LIFE SCIENCES

Synonymer: Ro 31-8220 mesylate

BMLEI2830001
ENZOBMLEI2830001EA 1400 NOK
ENZOBMLEI2830001
Bisindolylmaleimide IX methanesulphonate ≥98%
Bisindolylmaleimide IX methanesulphonate

Cell-permeable PKC inhibitor. Selective for PKC (IC50=10nM) over CaM kinase II (IC50=17µM) and PKA (IC50=0.9µM). Induces apoptosis in various cell lines.


  • PKC inhibitor


Red/orange solid.


Soluble in DMSO (10mg/ml)

Formel: C₂₅H₂₃N₅O₂S·CH₄O₃S
MW: 457,6·96,1 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD03788207
CAS-nummer: 138489-18-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a14ecb3fd8335054c7966b5ba07f1864d3fc1e4d&vid=c73a598b88510dd9b1174a5c3e1e06086e605540&prid=web&clid=SAPDAM

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR