Geldanamycin ≥98% Biotin

Leverandør: ENZO LIFE SCIENCES

Synonymer: (4E,6Z,8S,9S,10E,12S,13R,14S,16R)-13-Hydroxy-8,14,19-trimethoxy-4,10,12,16-tetramethyl-3,20,22-trioxo-2-azabicyclo[16.3.1]docosa-1(21),4,6,10,18-pentaen-9-yl carbamate

BMLEI3410001
ENZOBMLEI3410001EA 3350 NOK
ENZOBMLEI3410001
Geldanamycin ≥98% Biotin
Geldanamycin

A useful tool for affinity purification of HSP-90 and HSP-90 dependent client proteins.


  • HSP90 inhibitor

Formel: C₂₉H₄₀N₂O₉
MDL nummer: MFCD00274570
CAS-nummer: 30562-34-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32b45bb0e5ddbf857980143d9e7b85cb1d2fa765&vid=6f0f481f8e03338983d73cf2d19294c43448e9b3&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR